
Zinc(II) wolframate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13597-56-3
:
Formula:O₄WZn
MDL Number:MFCD00049633
:313.22 g/mol
EINECS:237-053-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together