
4,5-Dichloro-3(2H)-pyridazinone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:932-22-9
:
Formula:C₄H₂Cl₂N₂O
MDL Number:MFCD00051504
:164.98 g/mol
Melting Pt:200…204 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together