Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
APOSOR24171-1G
:
Apollo Scientific
:
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
APOSOR24171-1G
:
Apollo Scientific
:
Pack type:
Glass bottle
Pk:
1 g
Seq:
3047
:
:
158.59 g/mol
:
27006-76-4
:
C₉H₁₁ClN₂O
:
MFCD00052538