Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Okadaic acid sodium salt 98%, high purity
  ACRO328400001
 :  
undefined
 
Okadaic acid sodium salt 98%, high purity
  ACRO328400001
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    0,1 mg
  • Seq:
    1286

 

 

  • Appearance:
    Clear white to off-white Solid or film
  • TLC:
    ≥97.5 %

 

 

 :
 :827.00 g/mol
 :Freezer
 :2811
 :6.1,II
 :GHS06
 :209266-80-8
 :78111-17-8
 :C₄₄H₆₇NaO₁₃
 :MFCD00210208