
Tri(2-furyl)phosphine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5518-52-5
:
Formula:C₁₂H₉O₃P
MDL Number:MFCD00151857
:232.17 g/mol
Boiling Pt:136 °C (4 mmHg)
Melting Pt:59…64 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together