Order Entry
Czech Republic
ContactUsLinkComponent
Tri(2-furyl)phosphine 98%
Irritant
Tri(2-furyl)phosphine 98%
Certificates
undefined
Irritant
Tri(2-furyl)phosphine 98%

Specification Test Results

  • Appearance:
    Off-white to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    61 to 64 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together