Order Entry
Czech Republic
ContactUsLinkComponent
1,5-Pentamethylene-1H-tetrazole
1,5-Pentamethylene-1H-tetrazole
Catalog # PROOMM0692.00
CAS Number:  
1,5-Pentamethylene-1H-tetrazole
Catalog # PROOMM0692.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :138.17 g/mol
EINECS:200-219-3
CAS Number:54-95-5
Formula:C₆H₁₀N₄
MDL Number:MFCD00005939