
1,5-Pentamethylene-1H-tetrazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54-95-5
:
Formula:C₆H₁₀N₄
MDL Number:MFCD00005939
:138.17 g/mol
Merck Index:14,7141
Boiling Pt:194 °C (12 mmHg)
Melting Pt:59…61 °C
Storage Temperature:Ambient
EINECS:200-219-3
ADR:6.1,II
:GHS06
Related Information
Frequently Bought Together