Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dicyclohexylzinc
Dicyclohexylzinc 0.4 mol/l in diethyl ether, Sigma-Aldrich®
Dicyclohexylzinc 0.4 mol/l in diethyl ether, Sigma-Aldrich®
SIAL718009-10ML
:
Merck
:
Dicyclohexylzinc 0.4 mol/l in diethyl ether, Sigma-Aldrich®
SIAL718009-10ML
:
Merck
:
Typ balení:
Glass bottle
Bal.:
10 mL
Sekvence:
2275
:
:
15658-08-9
:
C₁₂H₂₂Zn
:
MFCD16878978