1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:85-42-7
:
Formula:C₈H₁₀O₃
MDL Number:MFCD00064863
:154.17 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-604-9
Frequently Bought Together