Order Entry
Czech Republic
ContactUsLinkComponent
 
Buffers, tricine, 0,5 M solutions
 
Buffers, tricine, 0,5 M solutions
 

 

  • CAS No.:
    5704-04-1
  • MDL No.:
    MFCD00004277
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

Frequently Bought Together