Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
Buffer, imidazole buffer solution pH 7,5 (2 mol/L)
Buffer, imidazole buffer solution pH 7,5 (2 mol/L)
Buffer, imidazole buffer solution pH 7,5 (2 mol/L)
Catalog # 786-529
Supplier:
G-Biosciences
CAS Number:
Expand image
Buffer, imidazole buffer solution pH 7,5 (2 mol/L)
Catalog # 786-529
Supplier:
G-Biosciences
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Description:
Imidazole buffer solution pH 7,5 (2 mol/l)
Specifications
About this item
Product Details
Product Details