
Cerium(III) oxalate hydrate, Sigma-Aldrich®
Catalog # SIAL325511-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:544.29 g/mol (anhy)
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:15750-47-7
:880358-87-2
Formula:C₆Ce₂O₁₂·H₂O
MDL Number:MFCD00150139