
Methyl vanillate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3943-74-6
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00008438
:182.18 g/mol
Boiling Pt:285…287 °C
Melting Pt:62…66 °C
Storage Temperature:Ambient
EINECS:223-525-9
:GHS07
Frequently Bought Together