

Hydrogen chloride 1 M in ethyl acetate, AcroSeal®
Catalog # 428921000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.95 to 1.15 M
- Refractive index:1.5090 to 1.5130 (20°C, 589 nm)
- GC:≥88 %
Specification Test Results
About this item
:
:36.46 g/mol
Density:0.9 g/cm³
Flash Pt:–3 °C
Storage Temperature:Ambient
EINECS:231-595-7
UN:2924
ADR:3,II
:GHS05
:GHS02
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324