ContactUsLinkComponent
Cleanroom overalls with open legs, 8R013
Cleanroom overalls with open legs, 8R013
Supplier:  FRISTADS KANSAS
Certificates
undefined
Cleanroom overalls with open legs, 8R013
Supplier:  FRISTADS KANSAS
Specifications
  • Catalog No:
  • KWIN1006515002XL
  • KWIN1006519003XL
  • KWIN1006519002XL
  • KWIN100651900M
  • KWIN100651900L
  • KWIN100651500S
  • KWIN100651900XL
  • KWIN100651500M
  • KWIN100651500XS
  • KWIN100651500XL
  • KWIN100651900XS
  • KWIN100651900S
  • KWIN100651500L
  • KWIN1006515003XL
  • Model:
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Open legs, 8R013, XR50
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Weight:
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²
  • 110 g/m²