ContactUsLinkComponent
Crystallisation plates, CrystalStar™, 24-well CombiPlate™ and CrystalBridge™
Crystallisation plates, CrystalStar™, 24-well CombiPlate™ and CrystalBridge™
Catalog # 737-3508
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
undefined
Crystallisation plates, CrystalStar™, 24-well CombiPlate™ and CrystalBridge™
Catalog # 737-3508
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Description:
    ComboPlate™, universal platform, pre-greased
  • Seq:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    24
Specifications
About this item
The CombiPlate™ is a universal 24 well crystallisation plate.
  • Clear PS combined with a flat distortion free bottom offers excellent optical properties
  • A flattened raised ring around each well reduces the risk of cross contamination and makes it possible to seal the wells with silicone grease and coverslips (Ø18 mm) or VIEWseal™ sealer
  • Slightly raised lid protects the coverslips and sealer during transport and storage
  • For sitting drop experiments, CrystalBridge™ inserts fit exactly into the wells of the CombiPlate™