Chemical resistant overalls, Essen 5967
: SIOEN
With its double-sided PVC coated polyester fabric ('Chemtex') this overall is perfectly suited for intensive use in industrial environments. It is exceptionally strong and tear resistant and it will protect users from chemicals, oils and greases.
- Fixed hood with peak
- Zip closure under double flap with touch and close fastening and press studs
- Raglan sleeves
- Elasticated wind cuff in sleeves and ankles
- High frequency welded seams
Weight: 360 g/m²
: EN 14605 (Type 3), EN 14605 (Type 4), EN 343 (4-1-X), EN ISO 13688
- Catalog No:
- SIOE5967/J40/3XL
- SIOE5967/J40/1XL
- SIOE5967/J40/2XL
- SIOE5967/J40/S
- SIOE5967/J40/M
- SIOE5967/J40/L
- Model:
- Essen
- Essen
- Essen
- Essen
- Essen
- Essen
- Aplikace oblečení:
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Chemical protection, Oils, fats and fuels, Industrial
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Typ uzávěru:
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Hmotnost:
- 360 g/m²
- 360 g/m²
- 360 g/m²
- 360 g/m²
- 360 g/m²
- 360 g/m²
Frequently Bought Together