UHPLC columns, Pinnacle® DB Silica (USP L3)
Supplier: Restek
Certificates
About this item
Highly base-deactivated spherical silica manufactured by Restek. Useful for normal phase separations. Replaces Hypersil® BDS and Pinnacle® Amine.
Frequently Bought Together