ContactUsLinkComponent
Ductless fume hoods, Paramount®
Ductless fume hoods, Paramount®
Supplier:  LABCONCO
Certificates
undefined
Ductless fume hoods, Paramount®
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • LBCP6963623
  • LBCP6963521
  • 135-2023
  • Model:
  • Paramount®
  • Paramount®
  • Paramount®
  • Treated air volume:
  • 985 at 100 fpm
  • 370 at 100 fpm
  • 374 at 100 fpm
  • Lighting:
  • Fluorescent light, 24 W
  • Fluorescent light, 24 W
  • Fluorescent light, 24 W
  • Electrical:
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz