ContactUsLinkComponent
Handheld refractometers, Rhino
Handheld refractometers, Rhino
Supplier:  REICHERT
Certificates
undefined
Handheld refractometers, Rhino
Supplier:  REICHERT
Specifications
  • Catalog No:
  • 635-2131
  • 635-2130
  • 635-2132
  • Temperature compensation range:
  • −18…+40 °C
  • −18…+40 °C
  • −18…+40 °C
  • W×D×H:
  • 235×38×38 mm
  • 235×38×38 mm
  • 235×38×38 mm
  • Weight:
  • 162,4 g
  • 162,4 g
  • 162,4 g