ContactUsLinkComponent
Tubing cutter for polymer tubing, Upchurch Scientific®
Tubing cutter for polymer tubing, Upchurch Scientific®
Catalog # 554-3032
Tubing cutter for polymer tubing, Upchurch Scientific®
Catalog # 554-3032
Specifications
  • Description:
    Capillary polymer tubing cutter for 360 µm + 1/32" OD tubing
  • Pk:
    1
Specifications
About this item
Designed to cut capillary-sized polymer tubing.
The cutter makes clean, perpendicular cuts without collapsing thin walls. The included tubing sleeves are for cutting 360 µm O.D. polymer capillary tubing. Alternative sleeves are available for cutting 510 µm and ¹/₃₂" ext. Ø tubing. All tubing sleeves are 2" long and are made of FEP.
Delivery: Supplied with a set of 10 tubing sleeves along with five replacement blades.