Precolumn MicroFilters, Upchurch Scientific®
Dodavatel: UPCHURCH SCIENTIFIC
Certifikáty
O tomto produktu
Precolumn MicroFilters are specifically designed to directly connect into microbore or analytical columns.
Total theoretical void volume is only 0,5 μl (includes frit volume) and the PEEK tubing used in the assembly of these units has a 125 μm int.Ø, virtually eliminating any mixing of the sample with the mobile phase.
Frequently Bought Together