Tubing cutter for polymer tubing, Upchurch Scientific®
Catalog # 554-3024
Supplier: UPCHURCH SCIENTIFIC
Specifications
- Description:Standard polymer tubing cutter for 1/16" - 1/8" OD tubing, complete
- Pk:1
Specifications
About this item
Designed to cut polymer tubing ¹/₁₆" and ¹/₈" O.D.
Complete with guide holes to ensure precise cutting. The cutter is durable, reliable and easy to operate.
Delivery: Complete unit supplied with five replacement blades.