ContactUsLinkComponent
Handheld refractometers for wine
 :  Atago
undefined
Handheld refractometers for wine
 :  Atago
  • Catalog No:
  • 635-0278
  • 635-0279
  • 635-0277
  • 635-0157
  • 635-0282
  • 635-0281
  • Sample size:
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • 0,3 ml
  • Response time:
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • 3 seconds
  • Temperature compensation range:
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • Electrical:
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • Two AAA batteries
  • W×D×H:
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • 55×31×109 mm
  • Weight:
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
  • 100 g
Frequently Bought Together