Formamide treated Reag. Ph. Eur. 1039201
Catalog # 87838.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:45.04 g/mol
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941