Order Entry
Czech Republic
ContactUsLinkComponent
Vanadium(V) oxide solution in sulphuric acid Reag. Ph. Eur. 1034001
Corrosive
Irritant
Health
Vanadium(V) oxide solution in sulphuric acid Reag. Ph. Eur. 1034001
Catalog # 87828.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Health
Vanadium(V) oxide solution in sulphuric acid Reag. Ph. Eur. 1034001
Catalog # 87828.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :181.88 g/mol
 :GHS05
 :GHS07
 :GHS08
CAS Number:1314-62-1
Formula:O₅V₂
MDL Number:MFCD00011457