Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cupri-citric solution
Cupri-citric solution Reag. Ph. Eur. 1023100
Cupri-citric solution Reag. Ph. Eur. 1023100
Irritant
Environment
Cupri-citric solution Reag. Ph. Eur. 1023100
Catalog # 87812.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Irritant
Environment
Cupri-citric solution Reag. Ph. Eur. 1023100
Catalog # 87812.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
UN:
3082
ADR:
9,III
:
GHS07
:
GHS09
Product Details
Documentation