4-Hydroxy-4-methyl-2-pentanone
Catalog # LACH30967
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:116.16 g/mol
Merck Index:13,02983
Boiling Pt:166 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:204-626-7
UN:1148
ADR:3,III
REACH:01-2119473975-21
:GHS02
:GHS07
CAS Number:123-42-2
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471