Sodium thiosulphate 0.5 mol/l (0.5 N) in aqueous solution 0.5 mol/l (0.5 N) in aqueous solution
Catalog # LACH62026002M1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
:
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499