Order Entry
Czech Republic
ContactUsLinkComponent
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # LACH61026010P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # LACH61026010P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499