Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution
Catalog # LACH61009001P00001
Supplier:  Lach-ner S.R.O.
CAS Number:  
Sodium hydroxide 0.05 mol/l (0.05 N) in aqueous solution
Catalog # LACH61009001P00001
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548