Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Test Papers
Indicator Papers
pH-Indicator paper phenolphthalein
pH-Indicator paper phenolphthalein
pH-Indicator paper phenolphthalein
Catalog # LACH51011000P00001
Supplier:
Lach-ner S.R.O.
Expand image
pH-Indicator paper phenolphthalein
Catalog # LACH51011000P00001
Supplier:
Lach-ner S.R.O.
Specifications
Description:
pH-Indicator paper phenolphthalein, strips
Environmentally Preferable:
Pk:
100
Specifications
About this item
Product Details
Product Details