0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Test Papers
pH Indicator Papers and Test Strips
pH-Indicator paper strips, Congo red
pH-Indicator paper strips, Congo red
LACH51012000P00001
:
Lach-ner S.R.O.
pH-Indicator paper strips, Congo red
LACH51012000P00001
:
Lach-ner S.R.O.
Description:
pH-Indicator paper strips, Congo red
Environmentally Preferable:
Pk:
100