
Fehling's reagent I (copper(II) sulphate solution)
Catalog # LACHFEHLING1
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:159.61 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3082
ADR:9,III
:GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981