Order Entry
Czech Republic
ContactUsLinkComponent
Cobalt(II) acetate tetrahydrate
Health
Environment
Cobalt(II) acetate tetrahydrate
Catalog # LACH40116AP0G0500
Supplier:  Lach-ner S.R.O.
CAS Number:  
Health
Environment
Cobalt(II) acetate tetrahydrate
Catalog # LACH40116AP0G0500
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :249.08 g/mol
Merck Index:12,02494
Melting Pt:140 °C
Density:1.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-755-8
UN:3077
ADR:9,III
 :GHS08
 :GHS09
CAS Number:6147-53-1
Formula:(H₃CCOO)₂Co·4H₂O
MDL Number:MFCD00149569