Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Morpholino)ethyl isothiocyanate
2-(4-Morpholino)ethyl isothiocyanate ≥97%
2-(4-Morpholino)ethyl isothiocyanate ≥97%
L12046.04
:
Thermo Fisher Scientific
:
2-(4-Morpholino)ethyl isothiocyanate ≥97%
L12046.04
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
2 g
Seq:
3107
:
:
162…163 °C (10 mmHg)
:
1.110
:
>110 °C (230 °F)
:
8,III
:
172.25 g/mol
:
GHS06
:
GHS05
:
63224-35-1
:
C₇H₁₂N₂OS
:
MFCD00041221