2-Naphthol
LACH40106CP0G0250
:
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
:
:12,06471
:286 °C (1013 hPa)
:121.6 °C
:1.217 g/cm³ (20 °C)
:153 °C
:Ambient
:205-182-7
:3077
:9,III
:144.17 g/mol
:GHS07
:GHS09
:135-19-3
:C₁₀H₈O
:MFCD00004067