

1-Naphthol
Catalog # LACH40105CP0G0500
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:144.17 g/mol
Merck Index:13,06409
Boiling Pt:282 °C (1013 hPa)
Melting Pt:94 °C
Density:1.224 g/cm³ (20 °C)
Flash Pt:125 °C
Storage Temperature:Ambient
EINECS:201-969-4
UN:2811
ADR:6.1,III
:GHS05
:GHS07
CAS Number:90-15-3
Formula:C₁₀H₈O
MDL Number:MFCD00003930