Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium carbonate
Potassium carbonate, anhydrous, pure
Potassium carbonate, anhydrous, pure
LACH30212CP0G1000
:
Lach-ner S.R.O.
:
Potassium carbonate, anhydrous, pure
LACH30212CP0G1000
:
Lach-ner S.R.O.
:
Pack type:
Plastic bottle for solids
Pk:
1 kg
Seq:
4024
:
:
138.21 g/mol
:
13,07702
:
891 °C
:
2.42 g/cm³ (20 °C)
:
209-529-3
:
GHS07
:
Ambient
:
584-08-7
:
K₂CO₃
:
MFCD00011382