Phenol
Glass bottle for solids, 100 g
MACH121-229-0100
: MACH
: Phenylalcohol, Carbolic acid, Hydroxybenzene
- :108-95-2
- :C₆H₅OH
- :MFCD00002143
- :94.11 g/mol
- :12,07390
- :181.7 °C (1013 hPa)
- :40.8 °C
- :1.0722 g/cm³ (20 °C)
- :81 °C
- :Ambient
- :203-632-7
- :1671
- :6.1,II
- :01-2119471329-32
- Pack type:Glass bottle for solids
- Pk:100 g