Diethyl ether stabilised
Glass bottle, 1 L
Catalog # LACH20018FT3M1000
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with phenidon
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



