Trisodium phosphate dodecahydrate
LACH30051AP0G1000
:
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3745
:
:380.12 g/mol
:13,08736
:∼75 °C
:1.62 g/cm³ (20 °C)
:Ambient
:231-509-8
:8,III
:01-2119489800-32-xxxx
:GHS07
:10101-89-0
:Na₃PO₄·12H₂O
:MFCD00149198