

Lead(II) acetate trihydrate
Catalog # LACH40118CP0G1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1.000 g
- Seq:3047
Specifications
About this item
:
:379.34 g/mol
Merck Index:13,05415
Boiling Pt:280 °C (1013 hPa)
Melting Pt:75 °C
Density:2.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-104-4
UN:1616
ADR:6.1,III
:GHS08
:GHS09
CAS Number:6080-56-4
Formula:(H₃CCOO)₂Pb·3H₂O
MDL Number:MFCD00150023