Order Entry
Czech Republic
ContactUsLinkComponent
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution
Catalog # LACH61020200P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution
Catalog # LACH61020200P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589