0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Hydrochloric acid
Hydrochloric acid 0.5 mol/l (0.5 N)
Hydrochloric acid 0.5 mol/l (0.5 N)
LACH61018002P0000
:
Lach-ner S.R.O.
:
Hydrochloric acid 0.5 mol/l (0.5 N)
LACH61018002P0000
:
Lach-ner S.R.O.
:
Pk:
1
Seq:
0001
:
:
8,II
:
36.46 g/mol
:
231-595-7
:
1789
:
GHS05
:
Ambient
:
7647-01-0
:
HCl
:
MFCD00011324