Order Entry
Czech Republic
ContactUsLinkComponent
 
Hydrochloric acid 0.2 mol/l (0.2 N)
Catalog # LACH61018005P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  
Hydrochloric acid 0.2 mol/l (0.2 N)
Catalog # LACH61018005P0000
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324