Order Entry
Czech Republic
ContactUsLinkComponent
 
Hydrochloric acid 0.1 mol/l (0.1 N)
  LACH61018010P0000
 :  
Hydrochloric acid 0.1 mol/l (0.1 N)
  LACH61018010P0000
 :  

 

  • Pk:
    1
  • Seq:
    0001

 

 

 :
 :1.004 g/cm³ (25 °C)
 :Ambient
 :231-595-7
 :8,III
 :36.46 g/mol
 :1789
 :7647-01-0
 :HCl
 :MFCD00011324