Order Entry
Czech Republic
ContactUsLinkComponent
Hydrofluoric acid 38 - 40%
Toxic
Corrosive
Hydrofluoric acid 38 - 40%
Catalog # LACH10024C40M1000
Supplier:  Lach-ner S.R.O.
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 38 - 40%
Catalog # LACH10024C40M1000
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS06
 :GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346