Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution
Catalog # LACH40046BT0M0250
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:692.54 g/mol
Density:1.01 g/cm³ (20 °C)
EINECS:238-676-6
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428